Invitrogen
Manufacturer Code:MA537603
Catalog # PIMA537603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 539CT14.1.1 |
Immunogen | KLH conjugated synthetic peptide between 1410-1438 amino acids from human COL3A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha1 (III) collagen; alpha1 (III) collagen AW550625 COL 3 COL3 COL3 alpha 1 COL3A1 Col3a-1 Collagen 3 collagen alpha 1 type III collagen alpha-1(III) chain collagen type III alpha 1 chain collagen fetal collagen type III alpha 1 collagen type III alpha 1 (Ehlers-Danlos syndrome type IV autosomal dominant) collagen type III alpha 1 chain fibril forming Collagen3 EDS4A Ehlers-Danlos syndrome type IV autosomal dominant FLJ34534 LOC100009177 MGC93704 minisatellite 10w detected by probe MMS10 mKIAA4231 MMS10-W Ms10w pro alpha 1 type III collagen procollagen type III alpha 1; Collagen alpha-1(III) chain; collagen, fetal; collagen, type III, alpha 1; Ehlers-Danlos syndrome type IV, autosomal dominant
Gene Aliases: COL3A1; EDS4A
UniProt ID: (Human) P02461
Entrez Gene ID: (Human) 1281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.