Invitrogen
Manufacturer Code:PA595595
Catalog # PIPA595595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Collagen III/COL3A1 recombit protein (Position: D1222-E1455). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha1 (III) collagen; Collagen alpha-1(III) chain; Collagen alpha-1(III) chain COL3A1; collagen, fetal; collagen, type III, alpha 1; Ehlers-Danlos syndrome type IV, autosomal dominant; minisatellite 10w detected by probe MMS10; procollagen, type III, alpha 1
Gene Aliases: AW550625; Col3a-1; COL3A1; EDS4A; mKIAA4231; MMS10-W; Ms10w
UniProt ID: (Human) P02461, (Mouse) P08121, (Rat) P13941
Entrez Gene ID: (Human) 1281, (Mouse) 12825, (Rat) 84032
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.