Invitrogen
Manufacturer Code:PA136000
Catalog # 01-672-202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Purified native collagen type III extracted from human skin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha1 (III) collagen; Collagen alpha-1(III) chain; collagen, fetal; collagen, type III, alpha 1; EDS4A FLJ34534; Ehlers-Danlos syndrome type IV, autosomal dominant
Gene Aliases: COL3A1; EDS4A
UniProt ID: (Human) P02461
Entrez Gene ID: (Human) 1281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.