Novus Biologicals
Manufacturer Code:NBP105169
Catalog # NBP105169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | Recombinant expressed PIIANP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1 type II collagen; Alpha-1 type II collagen ANFH AOM cartilage collagen chondrocalcin COL11A3 Collagen 2 collagen alpha-1(II) chain collagen II alpha-1 polypeptide collagen type 2 collagen type II alpha 1 collagen type II alpha 1 (primary osteoarthritis spondyloepiphysealdysplasia congenital) Collagen-2 MGC131516 SEDC; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; Chondrocalcin; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen, type II, alpha 1
Gene Aliases: ANFH; AOM; COL11A3; COL2A1; SEDC; STL1
UniProt ID: (Human) P02458
Entrez Gene ID: (Human) 1280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.