Invitrogen
Manufacturer Code:PA585108
Catalog # PIPA585108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human Collagen II. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1 type II collagen; ANFH antibody AOM antibody COL11A3 antibody MGC131516 antibody SEDC antibody COL2A1 antibody cartilage collagen antibody collagen alpha-1(II) chain antibody chondrocalcin antibody collagen II alpha-1 polypeptide antibody alpha-1 type II collagen antibody collagen type II alpha 1 antibody; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; Chondrocalcin; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen, type II, alpha 1; disproportionate micromelia; Procollagen II alpha 1; procollagen, type II, alpha 1
Gene Aliases: ANFH; AOM; CG2A1A; COL11A3; Col2; Col2a; Col2a-1; COL2A1; COLLII; Del1; Dmm; Lpk; M100856; Rgsc856; SEDC; STL1
UniProt ID: (Human) P02458, (Rat) P05539, (Mouse) P28481
Entrez Gene ID: (Human) 1280, (Rat) 25412, (Mouse) 12824
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.