Invitrogen
Manufacturer Code:PA127397
Catalog # PIPA127397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Collagen type I purified from rat skin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles,Store at -20°C,avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1 type I collagen; Collagen alpha-1(I) chain; collagen, type 1, alpha 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; Type I Collagen Collagen 1; type I-alpha 1 collagen
Gene Aliases: Col1a1; COLIA1
UniProt ID: (Rat) P02454
Entrez Gene ID: (Rat) 29393
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.