Abnova
Manufacturer Code:PAB13496
Catalog # 89-116-705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified Col2a1 from rat cartilage. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with 0.5mL distilled water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against native Col2a1.
For Research Use Only
Protein Aliases: Alpha-1 type II collagen; CG2A1A; Chondrocalcin; Collagen alpha-1(II) chain; collagen type II (Col2A1) gene, enhancer region; COLLII; Procollagen II alpha 1; procollagen, type II, alpha 1
Gene Aliases: CG2A1A; Col2a1; COLLII
UniProt ID: (Rat) P05539
Entrez Gene ID: (Rat) 25412
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.