Invitrogen
Manufacturer Code:PA569975
Catalog # PIPA569975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of rat Coilin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLN80 p80-coilin coilin p80 p80; Coilin; coilin p80; nuclear phosphoprotein of Cajal body; p80; p80-coilin
Gene Aliases: C79982; CLN80; COIL; p80; p80-coilin; wu:fb08h11
UniProt ID: (Human) P38432
Entrez Gene ID: (Rabbit) 100343244, (Dog) 480564, (Horse) 100070720, (Bovine) 507580, (Human) 8161, (Guinea pig) 100727482, (Mouse) 12812, (Rat) 50998, (Zebrafish) 58024
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.