Invitrogen
Manufacturer Code:PA5117048
Catalog # PIPA5117048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 297-576 of human Coilin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C79982 Cln80 COIL coilin coilin p80 nuclear phosphoprotein of Cajal body p80 p80-coilin; Coilin; coilin p80; nuclear phosphoprotein of Cajal body; p80; p80-coilin
Gene Aliases: C79982; CLN80; COIL; p80; p80-coilin
UniProt ID: (Human) P38432
Entrez Gene ID: (Human) 8161, (Mouse) 12812, (Rat) 50998
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.