Novus Biologicals
Manufacturer Code:NBP154669
Catalog # NBP154669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to COIL(coilin) The peptide sequence was selected from the C terminal of COIL. Peptide sequence DYSLLPLLAAAPQVGEKIAFKLLELTSSYSPDVSDYKEGRILSHNPETQQ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLN80p80-coilin coilin coilin p80 p80; Coilin; coilin p80; p80; p80-coilin
Gene Aliases: CLN80; COIL; p80-coilin
UniProt ID: (Human) P38432
Entrez Gene ID: (Human) 8161
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.