Invitrogen
Manufacturer Code:MA517223
Catalog # PIMA517223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AF2C2 |
Immunogen | Recombit human protein purified from E.coli (His-cofilin) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa phosphoprotein; CFL CFL-1 18 kDa phosphoprotein non-muscle isoform cofilin-1 p18 COF1; cofilin 1 (non-muscle); Cofilin, non-muscle isoform; Cofilin-1; epididymis secretory protein Li 15; p18
Gene Aliases: AA959946; CFL; CFL1; Cof; cofilin; HEL-S-15; n-cofilin
UniProt ID: (Human) P23528, (Rat) P45592, (Mouse) P18760
Entrez Gene ID: (Human) 1072, (Rat) 29271, (Mouse) 12631
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.