Novus Biologicals
Manufacturer Code:NB100818660.025ML
Catalog # NB017828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around aa. 1~5 (M-A-S-G-V) derived from Human cofilin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa phosphoprotein; 18 kDa phosphoprotein CFL cofilin 1 (non-muscle) Cofilin non-muscle isoform cofilin-1 p18; cofilin 1 (non-muscle); Cofilin, non-muscle isoform; Cofilin-1; epididymis secretory protein Li 15; p18
Gene Aliases: CFL; CFL1; cofilin; HEL-S-15
UniProt ID: (Human) P23528
Entrez Gene ID: (Human) 1072
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.