Novus Biologicals
Manufacturer Code:NBP243608
Catalog # NBP243608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1318 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human COPD. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Archain; Archain archain 1 archain vesicle transport protein 1 coatomer delta subunit coatomer protein complex subunit delta coatomer protein delta-COP coatomer subunit delta COPD Delta-coat protein delta-COP; archain vesicle transport protein 1; coatomer delta subunit; coatomer protein complex, subunit delta; coatomer protein delta-COP; Coatomer subunit delta; Delta-coat protein; Delta-COP
Gene Aliases: ARCN1; COPD
UniProt ID: (Human) P48444
Entrez Gene ID: (Human) 372
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.