Novus Biologicals
Manufacturer Code:NBP174234
Catalog # NBP174234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to the C terminal of F11. Immunizing peptide sequence RHLCGGSIIGNQWILTAAHCFYGVESPKILRVYSGILNQSEIKEDTSFFG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coagulation factor XI; coagulation factor XI EC 3.4.21 EC 3.4.21.27 FXIPlasma thromboplastin antecedent MGC141891 PTA; Coagulation factor XIa heavy chain; Coagulation factor XIa light chain; FXI; Plasma thromboplastin antecedent; PTA
Gene Aliases: F11; FXI
UniProt ID: (Human) P03951
Entrez Gene ID: (Human) 2160
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.