Novus Biologicals
Manufacturer Code:NBP243703
Catalog # NBP243703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 268 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Factor X. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activated factor Xa heavy chain; Coagulation factor X; coagulation factor X EC 3.4.21 EC 3.4.21.6 factor Xa FX FXA prothrombinase Stuart factor Stuart-Prower factor; Factor X heavy chain; Factor X light chain; factor Xa; prothrombinase; Stuart factor; Stuart-Prower factor
Gene Aliases: F10; FX; FXA
UniProt ID: (Human) P00742
Entrez Gene ID: (Human) 2159
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.