Novus Biologicals
Manufacturer Code:NBP299698UV
Catalog # NB089051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Coagulation Factor X (NP_000495.1 Met 1-Lys 488). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activated factor Xa heavy chain; Coagulation factor X; coagulation factor X EC 3.4.21 EC 3.4.21.6 factor Xa FX FXA prothrombinase Stuart factor Stuart-Prower factor; Factor X heavy chain; Factor X light chain; factor Xa; prothrombinase; Stuart factor; Stuart-Prower factor
Gene Aliases: F10; FX; FXA
UniProt ID: (Human) P00742
Entrez Gene ID: (Human) 2159
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.