Novus Biologicals
Manufacturer Code:NBP145150
Catalog # NBP145150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | RFF-VII/1 |
Immunogen | Purified human factor VII |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); coagulation factor VII coagulation factor VII (serum prothrombin conversion accelerator) EC 3.4.21 EC 3.4.21.21 Eptacog alfa FVII coagulation protein Proconvertin Serum prothrombin conversion accelerator SPCA; Eptacog alfa; Factor VII heavy chain; Factor VII light chain; FVII coagulation protein; Proconvertin; Serum prothrombin conversion accelerator; SPCA
Gene Aliases: F7; SPCA
UniProt ID: (Human) P08709
Entrez Gene ID: (Human) 2155
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.