Novus Biologicals
Manufacturer Code:NBP290180AF532
Catalog # NB070693
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Coagulation Factor III/Tissue Factor (P13726 Met1-Glu251). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD142; CD142 CD142 antigen Coagulation factor III coagulation factor III (thromboplastin tissue factor) FLJ17960 TF TFA Thromboplastin tissue factor; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); TF; Thromboplastin; Tissue factor
Gene Aliases: CD142; F3; TF; TFA
UniProt ID: (Human) P13726
Entrez Gene ID: (Human) 2152
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.