Novus Biologicals
Manufacturer Code:NBP2413810.025MG
Catalog # NB019688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 22 |
Immunogen | Thrombin-antithrombin (TAT) complexes |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation peptide fragment 1; Activation peptide fragment 2; Coagulation factor II; coagulation factor II (thrombin); Coagulation Factor II coagulation factor II (thrombin) receptor-like 2 Coagulation factor II receptor-like 2 Coagulation factor II receptor-like 2 (protease-actovated receptor 3) PAR-3 PAR3proteinase-activated receptor 3 protease-activated receptor 3 proteinase-activated receptor-3 PT serine protease Thrombin receptor-like 2; prepro-coagulation factor II; Prothrombin; prothrombin B-chain; serine protease; Thrombin heavy chain; Thrombin light chain
Gene Aliases: F2; PT; RPRGL2; THPH1
UniProt ID: (Human) P00734
Entrez Gene ID: (Human) 2147
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.