Abnova
Manufacturer Code:MAB11907
Catalog # 89-110-043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S65-35 |
Immunogen | Recombinant fusion protein corresponding to amino acids of 1308-1381 at cytoplasmic domain of rat Cntnap1. |
Conjugate | Streptavidin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant Cntnap1.
For Research Use Only
Protein Aliases: Caspr; caspr1; Contactin-associated protein 1; neurexin 4; Neurexin IV; Neurexin-4; p190; Paranodin
Gene Aliases: Caspr; Cntnap1; Nrxn4
UniProt ID: (Rat) P97846
Entrez Gene ID: (Rat) 84008
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.