Novus Biologicals
Manufacturer Code:NBP1975210.025MG
Catalog # NB018802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CLI-9 |
Immunogen | Purified human clusterin (TRPM-2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 40 40 sulfated glycoprotein 2 Aging-associated gene 4 protein aging-associated protein 4 apo-J APOJ Apolipoprotein J CLIclusterin (complement lysis inhibitor SP-40 clusterin Complement cytolysis inhibitor complement lysis inhibitor Complement-associated protein SP-40 Ku70-binding protein 1 KUB1SGP2 MGC24903 NA1/NA2 SGP-2 SP-40 sulfated glycoprotein 2 Testosterone-repressed prostate message 2 testosterone-repressed prostate message 2 apolipoprotein J) TRPM-2TRPM2; Aging-associated gene 4 protein; aging-associated protein 4; Apo-J; ApoJalpha; ApoJbeta; Apolipoprotein J; CLI; Clusterin; Clusterin alpha chain; Clusterin beta chain; Complement cytolysis inhibitor; Complement cytolysis inhibitor a chain; Complement cytolysis inhibitor b chain; complement lysis inhibitor; Complement-associated protein SP-40,40; Ku70-binding protein 1; NA1/NA2; SGP-2; Sulfated glycoprotein 2; Testosterone-repressed prostate message 2; TRPM-2
Gene Aliases: AAG4; APO-J; APOJ; CLI; CLU; CLU1; CLU2; KUB1; NA1/NA2; SGP-2; SGP2; SP-40; TRPM-2; TRPM2
UniProt ID: (Human) P10909
Entrez Gene ID: (Human) 1191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.