Novus Biologicals
Manufacturer Code:NBP1683080.05MG
Catalog # NBP16830805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The antiserum was produced against synthesized peptide derived from human CLU. The immunogen region is: C-terminal. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
For Research Use Only
Protein Aliases: Aging-associated gene 4 protein; aging-associated protein 4; Apo-J; ApoJalpha; ApoJbeta; Apolipoprotein J; CLI; Clusterin; Clusterin alpha chain; Clusterin beta chain; Complement cytolysis inhibitor; Complement cytolysis inhibitor a chain; Complement cytolysis inhibitor b chain; complement lysis inhibitor; Complement-associated protein SP-40,40; Ku70-binding protein 1; NA1/NA2; SGP-2; Sulfated glycoprotein 2; Testosterone-repressed prostate message 2; TRPM-2
Gene Aliases: AAG4; APO-J; APOJ; CLI; CLU; CLU1; CLU2; KUB1; NA1/NA2; SGP-2; SGP2; SP-40; TRPM-2; TRPM2
UniProt ID: (Human) P10909
Entrez Gene ID: (Human) 1191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.