Novus Biologicals
Manufacturer Code:NB10057531
Catalog # NB10057531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 527 and 577 of human cleavage stimulation factor 3' pre-RNA subunit 2 64kDa using the numbering given in entry NP_001316.1 (GeneID 1478). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: betaCstF-64 variant 2; betaCstF-64 variant 2 CF-1 64 kDa subunit Cleavage stimulation factor 64 kDa subunit cleavage stimulation factor subunit 2 cleavage stimulation factor 3' pre-RNA subunit 2 64kD cleavage stimulation factor 3' pre-RNA subunit 2 64kDa CSTF 64 kDa subunit CstF-64; CF-1 64 kDa subunit; Cleavage stimulation factor 64 kDa subunit; Cleavage stimulation factor subunit 2; cleavage stimulation factor, 3' pre-RNA, subunit 2; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kD; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa; CSTF 64 kDa subunit
Gene Aliases: CstF-64; CSTF2
UniProt ID: (Human) P33240
Entrez Gene ID: (Human) 1478
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.