Abnova
Manufacturer Code:MAB18316
Catalog # 89-338-241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S258-5 |
Immunogen | A synthetic peptide corresponding to amino acids 98-115 at N-terminus of rat Clcn3. |
Conjugate | ATTO680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: chloride channel 3; Chloride channel protein 3; Chloride transporter ClC-3; ClC-3; H(+)/Cl(-) exchange transporter 3; protein kinase C-regulated chloride channel
Gene Aliases: ClC-3; Clcn3
UniProt ID: (Rat) P51792
Entrez Gene ID: (Rat) 84360
Molecular Function: anion channel ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.