Proteintech
Manufacturer Code:164561AP150UL
Catalog # 50-173-0956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Claudin 3 Fusion Protein Ag9411 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C7orf1 claudin 3 Claudin3 CLDN3 CPE R 2 CPE R2 CPE receptor 2 CPETR2 HRVP1 RVP1; Claudin-3; Clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-receptor 2; hRVP1; Rat ventral prostate.1 protein; Rat ventral prostate.1 protein homolog; RVP1; ventral prostate.1 protein; ventral prostate.1 protein homolog; ventral prostate.1-like protein
Gene Aliases: AI182374; C7orf1; CLDN3; CPE-R2; CPETR2; HRVP1; mRVP1; RVP1
UniProt ID: (Human) O15551, (Rat) Q63400, (Mouse) Q9Z0G9
Entrez Gene ID: (Human) 1365, (Rat) 65130, (Mouse) 12739
Molecular Function: cell junction protein tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.