Invitrogen
Manufacturer Code:PA520754
Catalog # PIPA520754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 20 amino acid peptide near the carboxy terminus of human CLDN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Claudin-1; Senescence-associated epithelial membrane protein; senescence-associated epithelial membrane protein 1; UNQ481/PRO944 CLD1 ILVASC SEMP1 claudin-1 senescence-associated epithelial membrane protein 1
Gene Aliases: AI596271; CLD1; CLDN1; ILVASC; SEMP1; UNQ481/PRO944
UniProt ID: (Human) O95832, (Rat) P56745, (Mouse) O88551
Entrez Gene ID: (Dog) 608207, (Human) 9076, (Rat) 65129, (Mouse) 12737
Molecular Function: cell junction protein tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.