Novus Biologicals
Manufacturer Code:NB300613UV
Catalog # NB300613UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | X22 |
Immunogen | Purified human brain clathrin heavy chain. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CHC CHC17 clathrin heavy chain 1 Clathrin heavy chain on chromosome 17 clathrin heavy chain clathrin heavy chain (Hc) clathrin heavy polypeptide (Hc) clathrin heavy polypeptide-like 2 CLH17 CLH-17 CLTCL2 Hc KIAA0034; Clathrin heavy chain 1; Clathrin heavy chain on chromosome 17; clathrin, heavy chain (Hc); clathrin, heavy polypeptide (Hc); clathrin, heavy polypeptide-like 2; CLH-17
Gene Aliases: CHC; CHC17; CLH-17; CLH17; CLTC; CLTCL2; Hc; KIAA0034
UniProt ID: (Human) Q00610
Entrez Gene ID: (Human) 1213
Molecular Function: membrane traffic protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.