R&D Systems
Manufacturer Code:AF6784
Catalog # AF6784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Clathrin Heavy Chain 1/CHC17 Trp1521-Met1675 Accession # Q00610 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CHC CHC17 clathrin heavy chain 1 Clathrin heavy chain on chromosome 17 clathrin heavy chain clathrin heavy chain (Hc) clathrin heavy polypeptide (Hc) clathrin heavy polypeptide-like 2 CLH17 CLH-17 CLTC CLTCL2 Hc KIAA0034; Clathrin heavy chain 1; Clathrin heavy chain on chromosome 17; clathrin, heavy chain (Hc); clathrin, heavy polypeptide (Hc); clathrin, heavy polypeptide-like 2; CLH-17
Gene Aliases: CHC; CHC17; CLH-17; CLH17; CLTC; CLTCL2; Hc; KIAA0034
UniProt ID: (Human) Q00610
Entrez Gene ID: (Human) 1213
Molecular Function: membrane traffic protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.