Novus Biologicals
Manufacturer Code:NB100664130.1ML
Catalog # NB017771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Purified yeast cytosine deaminase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CDDCytidine aminohydrolase cytidine deaminase cytosine nucleoside deaminase EC 3.5.4.5 small cytidine deaminase; Cytidine aminohydrolase; Cytidine deaminase; cytosine nucleoside deaminase; small cytidine deaminase
Gene Aliases: CDA; CDD
UniProt ID: (Human) P32320
Entrez Gene ID: (Human) 978
Molecular Function: deaminase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.