R&D Systems
Manufacturer Code:MAB5005SP
Catalog # MAB5005SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 477226 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Cited-2 Met1-Cys269 Accession # O35740 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Cbp/p300-interacting transactivator 2; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain2cbp/p300-interacting transactivator 2 Cited2 MRG-1 MRG1melanocyte-specific gene 1-related gene 1 MSG1-related gene 1 MSG-related protein 1 P35srj; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 2; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2; melanocyte-specific gene 1-related gene 1; MRG-1; MSG-related protein 1; MSG1-related gene 1; P35srj
Gene Aliases: ASD8; CITED2; MRG-1; MRG1; P35SRJ; VSD2
UniProt ID: (Human) Q99967
Entrez Gene ID: (Human) 10370
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.