Abnova
Manufacturer Code:PAB15794
Catalog # 89-117-857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 191 amino acids of mouse Chtf18. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Chtf18.
For Research Use Only
Protein Aliases: 6030457M03Rik; Chl12; Chl12 homolog; Chromosome transmission fidelity protein 18 homolog; CTF18, chromosome transmission fidelity factor 18 homolog; MGC38008
Gene Aliases: 6030457M03Rik; Chl12; Chtf18; CTF18
UniProt ID: (Mouse) Q8BIW9
Entrez Gene ID: (Mouse) 214901
Molecular Function: DNA binding protein hydrolase nucleic acid binding nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.