Novus Biologicals
Manufacturer Code:NBP128466
Catalog # NBP128466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KGETLSQEEQRKLEED corresponding to internal region according to NP_005189.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CHETKcholine/ethanolamine kinase beta CHKLcholine kinase-like choline kinase betaEK Choline kinase-like protein CK CKB CKEKBEKBcholine/ethanolamine kinase EC 2.7.1.32 EC 2.7.1.82 Ethanolamine kinase Ethanolamine kinase beta; Choline kinase beta; Choline kinase-like protein; Choline/ethanolamine kinase; choline/ethanolamine kinase beta; CK; CKEKB; EK; EKB; Ethanolamine kinase; Ethanolamine kinase beta
Gene Aliases: CHETK; CHKB; CHKL; CK; CKB; CKEKB; EK; EKB; MDCMC
UniProt ID: (Human) Q9Y259
Entrez Gene ID: (Human) 1120
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.