Novus Biologicals
Manufacturer Code:NBP29217120UL
Catalog # NB026234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 240-335 of human CCKBR (NP_795344.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-B receptor; CCK-BR; CCK2 receptor; CCK2 receptor CCK2R CCK2-R CCK-B CCK-B receptor CCK-BR CCKRB cholecystokinin B receptor Cholecystokinin-2 receptor cholecystokinin-B receptor/gastrin receptor GASR gastrin receptor gastrin/cholecystokinin type B receptor; CCK2-R; Cholecystokinin-2 receptor; gastrin receptor; Gastrin/cholecystokinin type B receptor
Gene Aliases: CCK-B; CCK2R; CCKBR; CCKRB; GASR
UniProt ID: (Human) P32239
Entrez Gene ID: (Human) 887
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.