Novus Biologicals
Manufacturer Code:NBP267541
Catalog # NBP267541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ST57-09 |
Immunogen | C terminal Human Chk1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Cell cycle checkpoint kinase; Checkpoint kinase-1; Checkpoint S. pombe homolog of 1 CHK1 (checkpoint S.pombe) homolog CHK1 checkpoint homolog (S. pombe) CHK1serine/threonine-protein kinase Chk1 EC 2.7.11 EC 2.7.11.1; Checkpoint, S. pombe, homolog of, 1; CHK1 checkpoint homolog; Chk1-S; Serine/threonine-protein kinase Chk1
Gene Aliases: CHEK1; CHK1
UniProt ID: (Human) O14757
Entrez Gene ID: (Human) 1111
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.