Novus Biologicals
Manufacturer Code:NB100464
Catalog # NB100464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 250 and 300 of human Checkpoint Kinase 1 using the numbering given in SwissProt entry O14757 (GeneID 1111). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle checkpoint kinase; Checkpoint kinase-1; Checkpoint S. pombe homolog of 1 CHK1 (checkpoint S.pombe) homolog CHK1 checkpoint homolog (S. pombe) CHK1serine/threonine-protein kinase Chk1 EC 2.7.11 EC 2.7.11.1; Checkpoint, S. pombe, homolog of, 1; CHK1 checkpoint homolog; Chk1-S; Serine/threonine-protein kinase Chk1
Gene Aliases: CHEK1; CHK1
UniProt ID: (Human) O14757
Entrez Gene ID: (Human) 1111
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.