R&D Systems
Manufacturer Code:AF1630
Catalog # AF1630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Chk1 Tyr157-Thr476 Accession # O14757 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle checkpoint kinase; Checkpoint kinase-1; Checkpoint S. pombe homolog of 1 CHEK1 CHK1 (checkpoint S.pombe) homolog CHK1 checkpoint homolog (S. pombe) CHK1serine/threonine-protein kinase Chk1 EC 2.7.11 EC 2.7.11.1; Checkpoint, S. pombe, homolog of, 1; CHK1 checkpoint homolog; Chk1-S; Serine/threonine-protein kinase Chk1
Gene Aliases: CHEK1; CHK1
UniProt ID: (Human) O14757
Entrez Gene ID: (Human) 1111
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.