Novus Biologicals
Manufacturer Code:NBP197774
Catalog # NBP197774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence EHGPSPGQDTFCQGK corresponding to internal region (near C-Terminus) according to NP_003456.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CHITCHI3 chitinase 1 (chitotriosidase) Chitinase-1 chitotriosidase-1 EC 3.2.1.14 FLJ00314 MGC125322 plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase; chitinase 1 (chitotriosidase); Chitinase-1; Chitotriosidase-1; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase
Gene Aliases: CHI3; CHIT; CHIT1; CHITD
UniProt ID: (Human) Q13231
Entrez Gene ID: (Human) 1118
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.