Invitrogen
Manufacturer Code:PA547720
Catalog # PIPA547720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Chitotriosidase/CHIT1 Ala22-Ser464 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CHI3 CHIT CHITD chitinase-1 chitotriosidase-1 plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase; Chitinase-1; Chitotriosidase-1
Gene Aliases: 2300002L19Rik; Chit1
UniProt ID: (Mouse) Q9D7Q1
Entrez Gene ID: (Mouse) 71884
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.