Novus Biologicals
Manufacturer Code:NBP221969AF405
Catalog # NP221969A40
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 9X24 |
Immunogen | This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a Rat immunized with human recombinant protein of Chitinase 3-like 1. Rat anti human Chitinase 3-like 1 NP_001267. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: 39 kDa synovial protein; AW208766 Brp39 Chi3l1 gp39; Cartilage glycoprotein 39; CGP-39; chitinase 3-like 1 (cartilage glycoprotein-39); Chitinase-3-like protein 1; YKL-40
Gene Aliases: ASRT7; CGP-39; CHI3L1; GP-39; GP39; HC-gp39; hCGP-39; HCGP-3P; YKL-40; YKL40; YYL-40
UniProt ID: (Human) P36222
Entrez Gene ID: (Human) 1116
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.