R&D Systems
Manufacturer Code:MAB2649
Catalog # MAB2649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 321924 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Chitinase 3-like 1 Tyr22-Ala381 Accession # Q61362 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 39 kDa synovial protein; AW208766 Brp39 Chi3l1 Chitinase 3 like 1 gp39 HCgp39 YKL-40; Cartilage glycoprotein 39; CGP-39; chitinase 3-like 1 (cartilage glycoprotein-39); Chitinase-3-like protein 1; YKL-40
Gene Aliases: ASRT7; CGP-39; CHI3L1; GP-39; GP39; HC-gp39; hCGP-39; HCGP-3P; YKL-40; YKL40; YYL-40
UniProt ID: (Human) P36222
Entrez Gene ID: (Human) 1116
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.