Novus Biologicals
Manufacturer Code:NBP128610AF350
Catalog # NP128610A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C10 |
Immunogen | Recombinant human CHI3L1 (22-383aa) purified from E. coli (NP_001267). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 39 kDa synovial protein; AW208766 Brp39 Chi3l1 gp39; Cartilage glycoprotein 39; CGP-39; chitinase 3-like 1 (cartilage glycoprotein-39); Chitinase-3-like protein 1; YKL-40
Gene Aliases: ASRT7; CGP-39; CHI3L1; GP-39; GP39; HC-gp39; hCGP-39; HCGP-3P; YKL-40; YKL40; YYL-40
UniProt ID: (Human) P36222
Entrez Gene ID: (Human) 1116
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.