Abnova
Manufacturer Code:H00055790B02P
Catalog # 89-002-851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ChGn (AAH60772.1 1 a.a. ~ 297 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ChGn protein.
For Research Use Only
Protein Aliases: beta4GalNAcT; Beta4GalNAcT-1; ChGn; Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 1; chondroitin beta1,4 N-acetylgalactosaminyltransferase; Chondroitin sulfate N-acetylgalactosaminyltransferase 1; CSGalNAcT-1; FLJ11264; FLJ13760; glucuronylgalactosylproteoglycan 4-beta-N- acetylgalactosaminyltransferase
Gene Aliases: beta4GalNAcT; CHGN; CSGalNAcT-1; CSGALNACT1; GALNACT1; UNQ656/PRO1287
UniProt ID: (Human) Q8TDX6
Entrez Gene ID: (Human) 55790
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.