Invitrogen
Manufacturer Code:MA531382
Catalog # PIMA531382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL3169 |
Immunogen | Recombit protein corresponding to Human CHAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acetyl CoA:choline O-acetyltransferase; CHOACTase; choline acetylase; choline acetyltransferase; Choline O-acetyltransferase
Gene Aliases: B230380D24Rik; CHAT; CHOACTASE; CMS1A; CMS1A2; CMS6
UniProt ID: (Human) P28329
Entrez Gene ID: (Human) 1103, (Rat) 290567, (Mouse) 12647
Molecular Function:
acetyltransferase
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.