Invitrogen
Manufacturer Code:PA19027
Catalog # 01-674-611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the C terminus amino acids CKEKATRPSQGHQP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: acetyl CoA:choline O-acetyltransferase; CHOACTase; choline acetylase; Choline O-acetyltransferase; CMS1A CMS1A2 choline acetyltransferase acetyl CoA choline O-acetyltransferase
Gene Aliases: CHAT; CHOACTASE; CMS1A; CMS1A2; CMS6
UniProt ID: (Human) P28329
Entrez Gene ID: (Human) 1103
Molecular Function:
acetyltransferase
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.