Invitrogen
Manufacturer Code:PA558412
Catalog # PIPA558412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Cerebral Protein 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cerebral protein 1; Nucleomethylin; Ribosomal RNA-processing protein 8; Ribosomal RNA-processing protein 8 Cerebral protein 1 Nucleomethylin RRP8 KIAA0409 NML hucep-1 HUCE1 hucep1 RRP-8; RRP8 methyltransferase homolog
Gene Aliases: hucep-1; KIAA0409; NML; RRP8
UniProt ID: (Human) O43159
Entrez Gene ID: (Human) 23378
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.