Novus Biologicals
Manufacturer Code:NBP299621AF700
Catalog # NB088691
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Cerberus 1 (AAH69371.1 Met1-Ala267). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cerberus; cerberus 1, cysteine knot superfamily, homolog; cerberus cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily) cerberus 1 cysteine knot superfamily homolog (Xenopus laevis) cerberus-related 1 Cerberus-related protein DAN domain family member 4 DAND4 MGC119894 MGC119895 MGC96951; cerberus-related 1; Cerberus-related protein; DAN domain family member 4
Gene Aliases: CER1; DAND4
UniProt ID: (Human) O95813
Entrez Gene ID: (Human) 9350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.