Novus Biologicals
Manufacturer Code:NB1002911AF350
Catalog # N102911A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of human Ceramide Kinase (between residues 50-150). [Swiss-ProtNo. Q8TCT0] |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: Acylsphingosine kinase; Ceramide kinase; ceramide kinase dA59H18.2 DKFZp434E0211 EC 2.7.1.138 FLJ21430 FLJ23239 hCERKLipid kinase 4 KIAA1646dA59H18.3 lipid kinase LK4 LK4Acylsphingosine kinase MGC131878; hCERK; Lipid kinase 4; lipid kinase LK4; LK4
Gene Aliases: CERK; dA59H18.2; dA59H18.3; hCERK; KIAA1646; LK4
UniProt ID: (Human) Q8TCT0
Entrez Gene ID: (Human) 64781
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.