Invitrogen
Manufacturer Code:PA589398
Catalog # PIPA589398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-381 of human ADAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arf-GAP with dual PH domain-containing protein 2; centaurin, alpha 2; Centaurin-alpha-2; Cnt-a2
Gene Aliases: Adap2; Centa2
UniProt ID: (Mouse) Q8R2V5
Entrez Gene ID: (Mouse) 216991
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.