Invitrogen
Manufacturer Code:PA518207
Catalog # PIPA518207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RLTASTESGRSSR) corresponding to the C-terminus amino acids of ADAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAP2 CENTA2 centaurin alpha 2 HSA272195 centaurin-alpha 2 protein ArfGAP with dual PH domains 2 cent-b centaurin beta; Arf-GAP with dual PH domain-containing protein 2; centaurin beta; centaurin, alpha 2; centaurin-alpha 2 protein; Centaurin-alpha-2; Cnt-a2
Gene Aliases: ADAP2; cent-b; CENTA2; HSA272195
UniProt ID: (Human) Q9NPF8
Entrez Gene ID: (Human) 55803
Molecular Function:
G-protein modulator
enzyme modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.