Novus Biologicals
Manufacturer Code:NBP29212320UL
Catalog # NB026139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human ADAP2 (NP_060874.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arf-GAP with dual PH domain-containing protein 2; arf-GAP with dual PH domain-containing protein 2 ArfGAP with dual PH domains 2 CENTA2centaurin-alpha 2 protein centaurin beta centaurin alpha 2 centaurin-alpha-2 cent-b Cnt-a2 HSA272195; centaurin beta; centaurin, alpha 2; centaurin-alpha 2 protein; Centaurin-alpha-2; Cnt-a2
Gene Aliases: ADAP2; cent-b; CENTA2; HSA272195
UniProt ID: (Human) Q9NPF8
Entrez Gene ID: (Human) 55803
Molecular Function:
G-protein modulator
enzyme modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.